missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylaniline, 99%, Thermo Scientific Chemicals

Código de producto. 11464050
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
1000 g
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

2,6-Dimethylaniline is used in pharmaceuticals, as dye intermediates and in organic synthesis. It is also used in the production of antioxidants, agricultural, pharmaceutical, rubber chemicals and other target organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethylaniline is used in pharmaceuticals, as dye intermediates and in organic synthesis. It is also used in the production of antioxidants, agricultural, pharmaceutical, rubber chemicals and other target organic molecules.

Solubility
Slightly miscible with water. Miscible with ethanol and diethyl ether.

Notes
It is sensitive to air and light. Incompatible withacids, acid chlorides, acid anhydrides, oxidizing agents, chloroformates and halogens.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 87-62-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007747
InChI Key UFFBMTHBGFGIHF-UHFFFAOYSA-N
Synonym 2,6-xylidine, 2-amino-m-xylene, o-xylidine, 2,6-dimethylbenzenamine, 2-amino-1,3-dimethylbenzene, 2,6-dimethylphenylamine, benzenamine, 2,6-dimethyl, 2,6-xylylamine, 2-amino-1,3-xylene, 1-amino-2,6-dimethylbenzene
PubChem CID 6896
ChEBI CHEBI:28738
IUPAC Name 2,6-dimethylaniline
SMILES CC1=CC=CC(C)=C1N

Especificaciones

Melting Point 11°C
Density 0.981
Boiling Point 216°C
Quantity 100 g
Odor Characteristic
Refractive Index 1.56
UN Number UN1711
Beilstein 636332
Merck Index 14,10084
Solubility Information Slightly miscible with water. Miscible with ethanol and diethyl ether.
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2,6-Dimethylaniline
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • 2,6-Dimethylaniline
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Otros peligros
  • MIXTURE LIST-Contains : 2,6-Dimethylaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'