missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethylbenzaldehyde, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
An aldehyde containing building block used for proteomics research
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
An aldehyde containing building block used for proteomics research
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
An aldehyde containing building block used for proteomics research
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Specifications
Melting Point | 27°C to 30°C (literature) |
Color | White to Yellow |
Boiling Point | 230°C (literature) |
Flash Point | 96°C (205°F) |
Refractive Index | 1.548 |
Quantity | 5 g |
Beilstein | 907019 |
Sensitivity | Air Sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 134.18 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'