missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylmorpholine, 97%, mixture of isomers, Thermo Scientific Chemicals

Product Code. 11916811
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-91-3
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
InChI Key HNVIQLPOGUDBSU-UHFFFAOYSA-N
Synonym morpholine, 2,6-dimethyl, 2,6-dimethyl morpholine, 2,6-dimethylmorfolin, 2,6-dimethylmorfolin czech, ccris 5910, trans-2,6-dimethylmorpholine, 2,6-dimethyl-2,3,5,6-tetrahydro-4h-1,4-oxazine, 2,6-dimethyl-morpholine, 2,6-dimethylmorpholine, cis + trans, acmc-209nod
PubChem CID 110862
IUPAC Name 2,6-dimethylmorpholine
SMILES CC1CNCC(O1)C

Specifications

Melting Point -85.0°C
Density 0.9350g/mL
Boiling Point 147.0°C
Flash Point 48°C
Assay Percent Range 96% min. (mixture of isomers) (GC)
Packaging Glass bottle
Refractive Index 1.447
Quantity 25 g
Specific Gravity 0.935
Formula Weight 115.17
Percent Purity 97%
Chemical Name or Material 2, 6-Dimethylmorpholine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,6-Dimethylmorpholine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'