Learn More
2,6-Dimethylphenol, 99%, Thermo Scientific Chemicals
£31.90 - £102.00
Chemical Identifiers
CAS | 576-26-1 |
---|---|
Molecular Formula | C8H10O |
Molecular Weight (g/mol) | 122.17 |
MDL Number | MFCD00002240 |
InChI Key | NXXYKOUNUYWIHA-UHFFFAOYSA-N |
Synonym | 2,6-xylenol, phenol, 2,6-dimethyl, 2,6-dimethyl phenol, 2-hydroxy-m-xylene, 1-hydroxy-2,6-dimethylbenzene, vic-m-xylenol, 2,6-dmp, 2,6-dimethyl-phenol, xylenol 235, 2-hydroxy-1,3-dimethylbenzene |
PubChem CID | 11335 |
IUPAC Name | 2,6-dimethylphenol |
SMILES | CC1=CC=CC(C)=C1O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11413954
|
Thermo Scientific Alfa Aesar
A11947.36 |
500 g |
£31.90
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423954
|
Thermo Scientific Alfa Aesar
A11947.0E |
2500 g |
£102.00
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,6-dimethylphenol is used in the synthesis of anti-oxidant compounds due to the phenol moiety in the structure. In addition, this compound is used as a reactant in the synthesis of polyphenylene ether polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-dimethylphenol is used in the synthesis of anti-oxidant compounds due to the phenol moiety in the structure. In addition, this compound is used as a reactant in the synthesis of polyphenylene ether polymers.
Solubility
Soluble in water and chloroform.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from strong bases and oxidizing agent.
Chemical Identifiers
576-26-1 | |
122.17 | |
NXXYKOUNUYWIHA-UHFFFAOYSA-N | |
11335 | |
CC1=CC=CC(C)=C1O |
C8H10O | |
MFCD00002240 | |
2,6-xylenol, phenol, 2,6-dimethyl, 2,6-dimethyl phenol, 2-hydroxy-m-xylene, 1-hydroxy-2,6-dimethylbenzene, vic-m-xylenol, 2,6-dmp, 2,6-dimethyl-phenol, xylenol 235, 2-hydroxy-1,3-dimethylbenzene | |
2,6-dimethylphenol |
Specifications
576-26-1 | |
1.15 | |
78°C (172°F) | |
MFCD00002240 | |
1446677 | |
14,10082 | |
Soluble in water and chloroform. | |
CC1=CC=CC(C)=C1O | |
122.17 | |
122.17 | |
2,6-Dimethylphenol |
44°C to 48°C | |
203°C | |
C8H10O | |
UN2261 | |
Light sensitive | |
2,6-xylenol, phenol, 2,6-dimethyl, 2,6-dimethyl phenol, 2-hydroxy-m-xylene, 1-hydroxy-2,6-dimethylbenzene, vic-m-xylenol, 2,6-dmp, 2,6-dimethyl-phenol, xylenol 235, 2-hydroxy-1,3-dimethylbenzene | |
NXXYKOUNUYWIHA-UHFFFAOYSA-N | |
2,6-dimethylphenol | |
11335 | |
99% |
Safety and Handling
GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H311-H314
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID
missing translation for 'einecsNumber' : 209-400-1
missing translation for 'rtecsNumber' : ZE6125000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only