Learn More
2,6-Dimethylphenol, 99%, Thermo Scientific Chemicals
£16.18 - £236.18
Chemical Identifiers
CAS | 576-26-1 |
---|---|
Molecular Formula | C8H10O |
Molecular Weight (g/mol) | 122.17 |
MDL Number | MFCD00002240 |
InChI Key | NXXYKOUNUYWIHA-UHFFFAOYSA-N |
Synonym | 2,6-xylenol, phenol, 2,6-dimethyl, 2,6-dimethyl phenol, 2-hydroxy-m-xylene, 1-hydroxy-2,6-dimethylbenzene, vic-m-xylenol, 2,6-dmp, 2,6-dimethyl-phenol, xylenol 235, 2-hydroxy-1,3-dimethylbenzene |
PubChem CID | 11335 |
IUPAC Name | 2,6-dimethylphenol |
SMILES | CC1=CC=CC(C)=C1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10447300
|
Thermo Scientific Acros
116570050 |
5 g | Glass bottle |
£16.18
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10447490
|
Thermo Scientific Acros
116572500 |
250 g | Glass bottle |
£21.91
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10050730
|
Thermo Scientific Acros
116570010 |
1 kg | Glass bottle |
£57.79
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10052823
|
Thermo Scientific Acros
116570025 |
2.5 kg | Glass bottle |
£119.60
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10540404
|
Thermo Scientific Acros
116570051 |
5 kg | Plastic Bottle |
£236.18
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
576-26-1 | |
122.17 | |
NXXYKOUNUYWIHA-UHFFFAOYSA-N | |
11335 | |
CC1=CC=CC(C)=C1O |
C8H10O | |
MFCD00002240 | |
2,6-xylenol, phenol, 2,6-dimethyl, 2,6-dimethyl phenol, 2-hydroxy-m-xylene, 1-hydroxy-2,6-dimethylbenzene, vic-m-xylenol, 2,6-dmp, 2,6-dimethyl-phenol, xylenol 235, 2-hydroxy-1,3-dimethylbenzene | |
2,6-dimethylphenol |
Specifications
576-26-1 | |
100.0 | |
Brown-Yellow to Colorless | |
203.0°C | |
Authentic | |
C8H10O | |
MFCD00002240 | |
15, 10279 | |
2,6-xylenol, phenol, 2,6-dimethyl, 2,6-dimethyl phenol, 2-hydroxy-m-xylene, 1-hydroxy-2,6-dimethylbenzene, vic-m-xylenol, 2,6-dmp, 2,6-dimethyl-phenol, xylenol 235, 2-hydroxy-1,3-dimethylbenzene | |
NXXYKOUNUYWIHA-UHFFFAOYSA-N | |
2,6-dimethylphenol | |
11335 | |
99% | |
2, 6-Dimethylphenol |
98.5 | |
45.0°C to 48.0°C | |
1.1500g/mL | |
73°C | |
98.5% min. (GC) | |
(CH3)2C6H3OH | |
06, 485 | |
1.15 | |
Solubility in water: 10g/L (20°C). Other solubilities: soluble in naoh solutions,freely soluble in alcohol,chloroform,ether and,benzene | |
CC1=CC=CC(C)=C1O | |
122.17 | |
122.17 | |
Low Melting Solid |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-400-1
Recommended Storage : this product gradually takes up color on storage
RUO – Research Use Only