missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific Chemicals

Product Code. 15400816
Click to view available options
Quantity:
100 mL

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 21450-64-6
Molecular Formula C8H9BrMg
MDL Number MFCD00192083
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Synonym 2,6-dimethylphenylmagnesium bromide, 2,6-dimethylphenylmagnesium bromide solution, biguvduzgajwqk-uhfffaoysa-m, 2,6-dimethylphenymagnesium bromide, bromo 2,6-dimethylphenyl magnesium, 2,6-dimethyl-phenylmagnesium bromide, 2,6-dimethylphenyl magnesium bromide, 2,6-dimethylphenylmagnesium bromide, 0.5m in 2-methf, 2,6-dimethylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 2,6-dimethylphenylmagnesium bromide solution, 1.0 m in thf, grignard reagent
InChI Key BIGUVDUZGAJWQK-UHFFFAOYSA-M
SMILES CC1=CC=CC(C)=C1[Mg]Br
IUPAC Name magnesium;1,3-dimethylbenzene-2-ide;bromide
Molecular Weight (g/mol) 209.37
PubChem CID 4517616
Formula Weight 209.37
Concentration or Composition (by Analyte or Components) 0.5M in 2-MeTHF
Chemical Name or Material 2,6-Dimethylphenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'