missing translation for 'onlineSavingsMsg'
Learn More

2,6-Lutidine, 99%

Product Code. 10686512 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10686512

Brand: Thermo Scientific Acros 125190010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 108-48-5 | C7H9N | 107.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-48-5
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00006345
InChI Key OISVCGZHLKNMSJ-UHFFFAOYSA-N
Synonym 2,6-lutidine, lutidine, pyridine, 2,6-dimethyl, 2,6-dimethypyridine, alpha,alpha'-lutidine, alpha,alpha'-dimethylpyridine, 2,6-dimethyl-pyridine, hsdb 79, unii-15fq5d0t3p, 2,6-lutidene
PubChem CID 7937
ChEBI CHEBI:32548
IUPAC Name 2,6-dimethylpyridine
SMILES CC1=CC=CC(C)=N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -6.0°C
Density 0.9200g/mL
Boiling Point 143.0°C to 145.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4966 to 1.4986
Quantity 1 L
Beilstein 20, 244
Fieser 01,626
Merck Index 15, 5677
Specific Gravity 0.92
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: soluble in alcohol and ether,miscible with dimethylformamide and,tetrahydrofuran
Formula Weight 107.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 6-Lutidine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,6-Lutidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 2,6-Lutidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.