missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Lutidine N-oxide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
2,6-Lutidine N-oxide is used as a ligand and form coordination complexes with metal salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,6-Lutidine N-oxide is used as a ligand and form coordination complexes with metal salts.
Solubility
Miscible with water.
Notes
Hygroscopic. Incompatible with oxidizing agents, moisture and water.
2,6-Lutidine N-oxide is used as a ligand and form coordination complexes with metal salts.
Solubility
Miscible with water.
Notes
Hygroscopic. Incompatible with oxidizing agents, moisture and water.
Specifications
Density | 1.102 |
Boiling Point | 108°C to 111°C (10 mmHg) |
Refractive Index | 1.5685 |
Quantity | 250 g |
Beilstein | 110551 |
Sensitivity | Hygroscopic |
Solubility Information | Miscible with water. |
Formula Weight | 123.16 |
Percent Purity | 98% |
Chemical Name or Material | 2,6-Lutidine N-oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'