missing translation for 'onlineSavingsMsg'
Learn More

2-(6-Methoxy-1-benzofuran-3-yl)acetic acid, 97%, Thermo Scientific™

Product Code. 10213182
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 69716-05-8
Molecular Formula C11H10O4
Molecular Weight (g/mol) 206.197
MDL Number MFCD02677734
InChI Key QCXJFLREQGIACT-UHFFFAOYSA-N
Synonym 2-6-methoxy-1-benzofuran-3-yl acetic acid, 2-6-methoxybenzofuran-3-yl acetic acid, 6-methoxy-1-benzofuran-3-yl acetic acid, 3-benzofuranacetic acid, 6-methoxy, 2-6-methoxybenzo b furan-3-yl acetic acid, maybridge3_004130, 4ddh, ms0, 6-methoxybenzofuran-3-acetic acid, 6-methoxybenzofuran-3-yl acetic acid
PubChem CID 736818
IUPAC Name 2-(6-methoxy-1-benzofuran-3-yl)acetic acid
SMILES COC1=CC2=C(C=C1)C(=CO2)CC(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 206.2
Percent Purity 97%
Chemical Name or Material 2-(6-Methoxy-1-benzofuran-3-yl)acetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'