Learn More
2,6-Pyridinedimethanol, 98%, Thermo Scientific Chemicals
£40.70 - £128.00
Chemical Identifiers
CAS | 1195-59-1 |
---|---|
Molecular Formula | C7H9NO2 |
Molecular Weight (g/mol) | 139.154 |
MDL Number | MFCD00006351 |
InChI Key | WWFMINHWJYHXHF-UHFFFAOYSA-N |
Synonym | 2,6-pyridinedimethanol, pyridine-2,6-diyldimethanol, 2,6-bis hydroxymethyl pyridine, 6-hydroxymethyl pyridin-2-yl methanol, pyridine-2,6-dimethanol, 2,6-dihydroxymethylpyridine, 2,6-bis-hydroxymethyl pyridine, 2,6-bis-hydroxymethyl-pyridine, 2,6-pyridinyl dimethanol, 6-hydroxymethyl-2-pyridyl methan-1-ol |
PubChem CID | 70957 |
IUPAC Name | [6-(hydroxymethyl)pyridin-2-yl]methanol |
SMILES | C1=CC(=NC(=C1)CO)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11377735
|
Thermo Scientific Alfa Aesar
L02527.06 |
5 g |
£40.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387735
|
Thermo Scientific Alfa Aesar
L02527.14 |
25 g |
£128.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,6-Pyridinedimethanol was used in the preparation of Fe(III) complexes [Fe(18)O(6)(OH)(8)(pdm)(10)(pdmH)(4)(H(2)O)(4)](ClO(4))(10) and Fe(9)O(4)(OH)(2)(O(2)CMe)(10)(pdm)(pdmH)(4)](NO(3)). It is a ligand used in the synthesis of a variety of metal complexes and catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Pyridinedimethanol was used in the preparation of Fe(III) complexes [Fe(18)O(6)(OH)(8)(pdm)(10)(pdmH)(4)(H(2)O)(4)](ClO(4))(10) and Fe(9)O(4)(OH)(2)(O(2)CMe)(10)(pdm)(pdmH)(4)](NO(3)). It is a ligand used in the synthesis of a variety of metal complexes and catalysts.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
1195-59-1 | |
139.154 | |
WWFMINHWJYHXHF-UHFFFAOYSA-N | |
70957 | |
C1=CC(=NC(=C1)CO)CO |
C7H9NO2 | |
MFCD00006351 | |
2,6-pyridinedimethanol, pyridine-2,6-diyldimethanol, 2,6-bis hydroxymethyl pyridine, 6-hydroxymethyl pyridin-2-yl methanol, pyridine-2,6-dimethanol, 2,6-dihydroxymethylpyridine, 2,6-bis-hydroxymethyl pyridine, 2,6-bis-hydroxymethyl-pyridine, 2,6-pyridinyl dimethanol, 6-hydroxymethyl-2-pyridyl methan-1-ol | |
[6-(hydroxymethyl)pyridin-2-yl]methanol |
Specifications
1195-59-1 | |
185°C (15 mmHg) | |
MFCD00006351 | |
2,6-pyridinedimethanol, pyridine-2,6-diyldimethanol, 2,6-bis hydroxymethyl pyridine, 6-hydroxymethyl pyridin-2-yl methanol, pyridine-2,6-dimethanol, 2,6-dihydroxymethylpyridine, 2,6-bis-hydroxymethyl pyridine, 2,6-bis-hydroxymethyl-pyridine, 2,6-pyridinyl dimethanol, 6-hydroxymethyl-2-pyridyl methan-1-ol | |
WWFMINHWJYHXHF-UHFFFAOYSA-N | |
[6-(hydroxymethyl)pyridin-2-yl]methanol | |
70957 | |
98% |
110°C to 114°C | |
C7H9NO2 | |
116016 | |
Soluble in water. | |
C1=CC(=NC(=C1)CO)CO | |
139.154 | |
139.15 | |
2,6-Pyridinedimethanol |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
missing translation for 'einecsNumber' : 214-803-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only