missing translation for 'onlineSavingsMsg'
Learn More

2-([7-(acetylamino)-6-(benzyloxy)-2-phenylperhydropyrano[3,2-d][1,3]dioxin-8-yl]oxy)propanoic acid, Thermo Scientific™

Product Code. 10490574
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 499104-69-7
Molecular Formula C25H29NO8
Molecular Weight (g/mol) 471.506
InChI Key JPPMVSNCFXDOJX-UHFFFAOYSA-N
Synonym 2-7-acetylamino-6-benzyloxy-2-phenylperhydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2r-2-4ar,6s,7r,8r,8as-7-acetamido-6-benzyloxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2-6-benzyloxy-7-acetamido-2-phenyl-hexahydro-2h-pyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, benzyl n-acetyl-4,6-o-benzylidenemuramic acid, 2-7-acetamido-6-benzyloxy-2-phenylhexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid, 2-7-acetamido-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano 3,2-d 1,3 dioxin-8-yl oxy propanoic acid
PubChem CID 3843768
IUPAC Name 2-[(7-acetamido-2-phenyl-6-phenylmethoxy-4,4a,6,7,8,8a-hexahydropyrano[3,2-d][1,3]dioxin-8-yl)oxy]propanoic acid
SMILES CC(C(=O)O)OC1C(C(OC2C1OC(OC2)C3=CC=CC=C3)OCC4=CC=CC=C4)NC(=O)C

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 471.5
Percent Purity 97%
Chemical Name or Material 2-{[7-(Acetylamino)-6-(benzyloxy)-2-phenylperhydropyrano[3, 2-d][1, 3]dioxin-8-yl]oxy}propanoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'