missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Dibromonaphthalene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is an intermediate in dyestuffs and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an intermediate in dyestuffs and pharmaceuticals.
Solubility
Soluble in acetonitrile.
Notes
Incompatible with oxidizing agents. Store at room temperature.
It is an intermediate in dyestuffs and pharmaceuticals.
Solubility
Soluble in acetonitrile.
Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications
Specifications
Melting Point | 139°C to 142°C |
Quantity | 250 mg |
UN Number | UN3077 |
Beilstein | 1862041 |
Solubility Information | Soluble in acetonitrile. |
Formula Weight | 285.97 |
Percent Purity | 99% |
Chemical Name or Material | 2,7-Dibromonaphthalene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'