missing translation for 'onlineSavingsMsg'
Learn More

2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane, 97%, Thermo Scientific Chemicals

Product Code. 10867661
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331465-71-5
Molecular Formula C18H39N4P
Molecular Weight (g/mol) 342.51
MDL Number MFCD03840347
InChI Key WFHPXSHLCFHEIA-UHFFFAOYSA-N
Synonym 2,8,9-triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo 3.3.3 undecane, triisobutylphosphatrane, 2,8,9-tris 2-methylpropyl-2,5,8,9-tetraaza-1-phosphabicyclo 3.3.3 undecane, acmc-1bmup, 4,6,11-tris 2-methylpropyl-1,4,6,11-tetraza-5-phosphabicyclo 3.3.3 undecane, nitrilotrisethylenetris isobutylimino phosphine, 2,8,9-tri-i-butyl-2,5,8,9-tetraaza-1-phosphabicyclo 3.3.3 undecane, 2,8,9-triisobutyl-2,5,8,9-tetraaza-1-phospha-bicyclo 3.3.3 undecane
PubChem CID 4571243
IUPAC Name 4,6,11-tris(2-methylpropyl)-1,4,6,11-tetraza-5-phosphabicyclo[3.3.3]undecane
SMILES CC(C)CN1CCN2CCN(P1N(CC2)CC(C)C)CC(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9640g/mL
Elemental Analysis 60.60 to 65.65% C, 11.02 to 11.94% H, 15.70 to 17.01% N
Boiling Point 110.0°C to 118.0°C (0.5 mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 10155
Specific Gravity 0.964
Solubility Information Solubility in water: insoluble
Formula Weight 342.51
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane
Show More Show Less
compliance-icons
Product Identifier
  • 2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo¢3.3.3!undecane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'