missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-bromobenzo[b]furan, 99%, Thermo Scientific™

Product Code. 11440080
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38220-75-6
Molecular Formula C10H7BrO2
Molecular Weight (g/mol) 239.068
MDL Number MFCD00276737
InChI Key YPIQPBGNNXXKNI-UHFFFAOYSA-N
Synonym 1-5-bromo-1-benzofuran-2-yl ethanone, 2-acetyl-5-bromobenzo b furan, 1-5-bromobenzofuran-2-yl ethanone, 1-5-bromo-1-benzofuran-2-yl ethan-1-one, 2-acetyl-5-bromobenzofuran, ethanone, 1-5-bromo-2-benzofuranyl, acmc-1ajfl, maybridge1_001462, 2-acetyl-5-bromo benzo b furan, ypiqpbgnnxxkni-uhfffaoysa
PubChem CID 705670
IUPAC Name 1-(5-bromo-1-benzofuran-2-yl)ethanone
SMILES CC(=O)C1=CC2=C(O1)C=CC(=C2)Br

Specifications

Quantity 5 g
Formula Weight 239.07
Percent Purity 99%
Chemical Name or Material 2-Acetyl-5-bromobenzo[b]furan
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'