missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylbenzoic acid, 99%, Thermo Scientific Chemicals

Product Code. 10122320
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-56-0
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00002475
InChI Key QDAWXRKTSATEOP-UHFFFAOYSA-N
Synonym o-acetylbenzoic acid, benzoic acid, 2-acetyl, 2-acetylbenzoicacid, acetophenone-2-carboxylic acid, 2-acetyl-benzoic acid, benzoic acid, acetyl, 3-hydroxy-3-methylphthalide, 2'-acetophenonecarboxylic acid, acetophenone-2'-carboxylic acid, methyl phenyl ketone-o-carboxylic acid
PubChem CID 68474
IUPAC Name 2-acetylbenzoic acid
SMILES CC(=O)C1=CC=CC=C1C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 115°C to 118°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3COC6H4COOH
Quantity 5 g
Beilstein 10, 690
Formula Weight 164.16
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Acetylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'