missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylthiophene, 98%, Thermo Scientific Chemicals

Product Code. 10467280
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-15-3
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.17
MDL Number MFCD00005442
InChI Key WYJOVVXUZNRJQY-UHFFFAOYSA-N
Synonym 2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone
PubChem CID 6920
IUPAC Name 1-thiophen-2-ylethanone
SMILES CC(=O)C1=CC=CS1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 10°C to 11°C
Color Yellow to Brown
Density 1.1600g/mL
Boiling Point 214°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C4H3S)COCH3
Refractive Index 1.5640 to 1.566
Quantity 100 g
Beilstein 17, 287
Specific Gravity 1.16
Solubility Information Solubility in water: 14g/L (30°C). Other solubilities: soluble in ethanol,benzene,chloroform,benzene
Formula Weight 126.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Acetylthiophene, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'