Learn More
2-Acetylthiophene, 98%, Thermo Scientific Chemicals
£36.60 - £120.00
Chemical Identifiers
CAS | 88-15-3 |
---|---|
Molecular Formula | C6H6OS |
Molecular Weight (g/mol) | 126.17 |
MDL Number | MFCD00005442 |
InChI Key | WYJOVVXUZNRJQY-UHFFFAOYSA-N |
Synonym | 2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone |
PubChem CID | 6920 |
IUPAC Name | 1-thiophen-2-ylethanone |
SMILES | CC(=O)C1=CC=CS1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10467280
|
Thermo Scientific Acros
102721000 |
100 g | Glass bottle |
£36.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10667893
|
Thermo Scientific Acros
102725000 |
500 g | Glass bottle |
£120.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
88-15-3 | |
126.17 | |
WYJOVVXUZNRJQY-UHFFFAOYSA-N | |
6920 | |
CC(=O)C1=CC=CS1 |
C6H6OS | |
MFCD00005442 | |
2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone | |
1-thiophen-2-ylethanone |
Specifications
88-15-3 | |
100.0 | |
Yellow to Brown | |
214°C | |
Authentic | |
C6H6OS | |
1.5640 to 1.566 | |
17, 287 | |
2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone | |
WYJOVVXUZNRJQY-UHFFFAOYSA-N | |
1-thiophen-2-ylethanone | |
6920 | |
98% | |
2-Acetylthiophene, 98% |
97.5 | |
10°C to 11°C | |
1.1600g/mL | |
91°C | |
97.5% min. (GC) | |
(C4H3S)COCH3 | |
MFCD00005442 | |
1.16 | |
Solubility in water: 14g/L (30°C). Other solubilities: soluble in ethanol,benzene,chloroform,benzene | |
CC(=O)C1=CC=CS1 | |
126.17 | |
126.17 | |
Liquid |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 201-804-6
missing translation for 'rtecsNumber' : OB6300000
missing translation for 'tsca' : TSCA
RUO – Research Use Only