missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetylthiophene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
2-Acetylthiophene is used as an intermediate in the manufacturing of drugs like Tiamonium Iodide, Suprofan, Stepronin, Tenonitrozole, Namirotene, etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Acetylthiophene is used as an intermediate in the manufacturing of drugs like Tiamonium Iodide, Suprofan, Stepronin, Tenonitrozole, Namirotene, etc.
Solubility
Insoluble in water.
Notes
It is sensitive to light. Incompatible with oxidizing agents.
2-Acetylthiophene is used as an intermediate in the manufacturing of drugs like Tiamonium Iodide, Suprofan, Stepronin, Tenonitrozole, Namirotene, etc.
Solubility
Insoluble in water.
Notes
It is sensitive to light. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 10°C to 12°C |
Density | 1.168 |
Boiling Point | 213°C to 214°C |
Flash Point | 91°C (195°F) |
Odor | Slightly Sweetish |
Refractive Index | 1.566 |
Quantity | 500 g |
UN Number | UN2810 |
Beilstein | 107910 |
Sensitivity | Light sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'