Learn More
2-Adamantanamine hydrochloride, 99%, Thermo Scientific Chemicals
£83.42 - £346.73
Chemical Identifiers
CAS | 10523-68-9 |
---|---|
Molecular Formula | C10H18ClN |
Molecular Weight (g/mol) | 187.71 |
MDL Number | MFCD00074743 |
InChI Key | WLDWDRZITJEWRJ-UHFFFAOYSA-N |
Synonym | 2-adamantanamine hydrochloride, adamantan-2-amine hydrochloride, 2-aminoadamantane hydrochloride, 2-adamantylamine hydrochloride, 2-adamantanamine hcl, 2-adamantanamine, hydrochloride, adamantan-2-ylamine hydrochloride, 1r,3r,5r,7r-adamantan-2-amine hydrochloride, tricyclo 3.3.1.13.7 dec-2-ylamine hydrochloride, tricyclo 3.3.1.13,7 decan-2-amine, hydrochloride |
PubChem CID | 25331 |
IUPAC Name | adamantan-2-amine;hydrochloride |
SMILES | [H+].[Cl-].NC1C2CC3CC(C2)CC1C3 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10215980
|
Thermo Scientific Acros
167300050 |
5 g | Glass bottle |
£83.42
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10530614
|
Thermo Scientific Acros
167300250 |
25 g | Glass bottle |
£346.73
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10523-68-9 | |
187.71 | |
WLDWDRZITJEWRJ-UHFFFAOYSA-N | |
25331 | |
[H+].[Cl-].NC1C2CC3CC(C2)CC1C3 |
C10H18ClN | |
MFCD00074743 | |
2-adamantanamine hydrochloride, adamantan-2-amine hydrochloride, 2-aminoadamantane hydrochloride, 2-adamantylamine hydrochloride, 2-adamantanamine hcl, 2-adamantanamine, hydrochloride, adamantan-2-ylamine hydrochloride, 1r,3r,5r,7r-adamantan-2-amine hydrochloride, tricyclo 3.3.1.13.7 dec-2-ylamine hydrochloride, tricyclo 3.3.1.13,7 decan-2-amine, hydrochloride | |
adamantan-2-amine;hydrochloride |
Specifications
10523-68-9 | |
100.0 | |
White | |
98.5% min. (GC) | |
MFCD00074743 | |
Solubility in water: soluble. Other solubilities: soluble in methanol and ethanol | |
[H+].[Cl-].NC1C2CC3CC(C2)CC1C3 | |
187.71 | |
187.71 | |
Crystalline Powder |
98.5 | |
>300.0°C | |
Authentic | |
C10H18ClN | |
2-adamantanamine hydrochloride, adamantan-2-amine hydrochloride, 2-aminoadamantane hydrochloride, 2-adamantylamine hydrochloride, 2-adamantanamine hcl, 2-adamantanamine, hydrochloride, adamantan-2-ylamine hydrochloride, 1r,3r,5r,7r-adamantan-2-amine hydrochloride, tricyclo 3.3.1.13.7 dec-2-ylamine hydrochloride, tricyclo 3.3.1.13,7 decan-2-amine, hydrochloride | |
WLDWDRZITJEWRJ-UHFFFAOYSA-N | |
adamantan-2-amine;hydrochloride | |
25331 | |
99% | |
2-Adamantanamine hydrochloride |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 234-074-2
- 2-Adamantanamine hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only