missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 15435385
Click to view available options
Quantity:
50 mL
Packaging:
ChemSeal™ bottles
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Adamantylzinc bromide used as a intermediate for pharmaceutical.

Solubility
Reacts with water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry
TRUSTED_SUSTAINABILITY

Specifications

CAS 171860-65-4
Color Black to Yellow
Density 1.018 g/mL
Packaging ChemSeal™ bottles
Molecular Formula C10H15BrZn
MDL Number MFCD01311401
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym 2-adamantylzinc bromide solution, adamantan-2-yl bromo zinc, adamantan-2-ide,bromozinc 1+, 2-adamantylzinc bromide,0.5m solution i, 2-adamantylzinc bromide solution, 0.5 m in thf, 2-adamantylzinc bromide 0.5 m in tetrahydrofuran, bromo 1r,3r,5r,7r-adamantan-2-yl zinc, 2-adamantylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
Solubility Information Reacts with water.
InChI Key UKVCFEOZGIQMNI-UHFFFAOYSA-M
SMILES C1C2CC3CC1CC(C2)[CH-]3.[Zn+]Br
IUPAC Name adamantan-2-ide;bromozinc(1+)
Molecular Weight (g/mol) 280.514
PubChem CID 4145216
Formula Weight 280.51
Physical Form Liquid
Chemical Name or Material 2-Adamantylzinc bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'