missing translation for 'onlineSavingsMsg'
Learn More

2-Allyl-L-glycine, 95%, Thermo Scientific™

Product Code. 15418627
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 195316-72-4
Molecular Formula C5H10ClNO2
Molecular Weight (g/mol) 151.59
MDL Number MFCD00002627
InChI Key DIDZZOWASZMNQW-WCCKRBBISA-N
Synonym s-2-aminopent-4-enoic acid hydrochloride, l-2-allylglycine hydrochloride, s---2-amino-4-pentenoic acid hydrochloride, 2s-2-aminopent-4-enoic acid hydrochloride, 4-pentenoic acid, 2-amino-, hydrochloride, 2s, l-allylglycine hydrochloride, l-alpha-allylglycine hydrochloride, s-2-amino-4-pentenoic acid hydrochloride, 4-pentenoic acid, 2-amino-, hydrochloride, s
PubChem CID 44630067
IUPAC Name (2S)-2-aminopent-4-enoic acid;hydrochloride
SMILES C=CCC(C(=O)O)N.Cl

Specifications

Quantity 250 mg
Formula Weight 115.13
Percent Purity 95%
Chemical Name or Material 2-Allyl-L-glycine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'