Learn More
2-Amino-1-methylbenzimidazole, 99+%, Thermo Scientific Chemicals
£80.00 - £222.00
Chemical Identifiers
CAS | 1622-57-7 |
---|---|
Molecular Formula | C8H9N3 |
Molecular Weight (g/mol) | 147.18 |
MDL Number | MFCD00142855 |
InChI Key | XDFZKQJLNGNJAN-UHFFFAOYSA-N |
Synonym | 2-amino-1-methylbenzimidazole, 1-methyl-1h-benzimidazol-2-amine, 1-methyl-1h-benzo d imidazol-2-amine, 1-methyl-2-aminobenzimidazole, 1h-benzimidazol-2-amine, 1-methyl, 1-methyl-1h-1,3-benzodiazol-2-amine, ccris 4355, 2-amino-1-methyl-benzimidazole, 1h-benzimidazol-2-amine,1-methyl, 1-methyl-1h-benzoimidazol-2-ylamine |
PubChem CID | 74187 |
IUPAC Name | 1-methylbenzimidazol-2-amine |
SMILES | CN1C(N)=NC2=CC=CC=C12 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10318910
|
Thermo Scientific Acros
276300025 |
2.5 g | Glass bottle |
£80.00
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10147713
|
Thermo Scientific Acros
276300100 |
10 g | Glass bottle |
£222.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1622-57-7 | |
147.18 | |
XDFZKQJLNGNJAN-UHFFFAOYSA-N | |
74187 | |
CN1C(N)=NC2=CC=CC=C12 |
C8H9N3 | |
MFCD00142855 | |
2-amino-1-methylbenzimidazole, 1-methyl-1h-benzimidazol-2-amine, 1-methyl-1h-benzo d imidazol-2-amine, 1-methyl-2-aminobenzimidazole, 1h-benzimidazol-2-amine, 1-methyl, 1-methyl-1h-1,3-benzodiazol-2-amine, ccris 4355, 2-amino-1-methyl-benzimidazole, 1h-benzimidazol-2-amine,1-methyl, 1-methyl-1h-benzoimidazol-2-ylamine | |
1-methylbenzimidazol-2-amine |
Specifications
1622-57-7 | |
100.0 | |
White to Beige | |
99% min. (GC) | |
MFCD00142855 | |
XDFZKQJLNGNJAN-UHFFFAOYSA-N | |
1-methylbenzimidazol-2-amine | |
74187 | |
99+% | |
2-Amino-1-methylbenzimidazole |
99.0 | |
203°C to 206°C | |
Authentic | |
C8H9N3 | |
2-amino-1-methylbenzimidazole, 1-methyl-1h-benzimidazol-2-amine, 1-methyl-1h-benzo d imidazol-2-amine, 1-methyl-2-aminobenzimidazole, 1h-benzimidazol-2-amine, 1-methyl, 1-methyl-1h-1,3-benzodiazol-2-amine, ccris 4355, 2-amino-1-methyl-benzimidazole, 1h-benzimidazol-2-amine,1-methyl, 1-methyl-1h-benzoimidazol-2-ylamine | |
CN1C(N)=NC2=CC=CC=C12 | |
147.18 | |
147.18 | |
Fine Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective glov
GHS Signal Word: Warning
RUO – Research Use Only