missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-thioxoethyl pivalate, 97%, Thermo Scientific™

Product Code. 10099601
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10099601

Brand: Thermo Scientific Maybridge RF02972EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175204-79-2
Molecular Formula C7H13NO2S
Molecular Weight (g/mol) 175.25
MDL Number MFCD00204238
InChI Key COULAOZTCJTHOX-UHFFFAOYSA-N
Synonym 2-amino-2-thioxoethyl pivalate, 2-amino-2-thioxoethyl 2,2-dimethylpropanoate, carbamothioylmethyl 2,2-dimethylpropanoate, 2-tert-butylcarbonyloxy thioacetamide, propanoicacid, 2,2-dimethyl-, 2-amino-2-thioxoethyl ester, propanoic acid, 2,2-dimethyl-, 2-amino-2-thioxoethyl ester, maybridge1_006303, 2-pivaloyloxy-thioacetamide, 2-amino-2-thioxoethylpivalate
PubChem CID 2782113
IUPAC Name (2-amino-2-sulfanylideneethyl) 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OCC(N)=S

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)3CCO2CH2C(S)NH2
Quantity 10 g
Formula Weight 175.25
Percent Purity 97%
Chemical Name or Material 2-Amino-2-thioxoethyl pivalate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.