missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dibromo-6-methylpyridine, 97%, Thermo Scientific Chemicals

Product Code. p-7025149
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11484497

Brand: Thermo Scientific Alfa Aesar B20498.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91872-10-5
Molecular Formula C6H6Br2N2
Molecular Weight (g/mol) 265.936
MDL Number MFCD00068229
InChI Key BLUYMDMBXIMIGK-UHFFFAOYSA-N
Synonym 2-amino-3,5-dibromo-6-methylpyridine, 3,5-dibromo-6-methyl-2-pyridinamine, 2-amino-3,5-dibromo-6-picoline, 6-amino-3,5-dibromo-2-picoline, 3,5-dibromo-6-methyl-pyridin-2-ylamine, 3,5-dibromo-6-methyl-2-pyridylamine, 2-pyridinamine, 3,5-dibromo-6-methyl, wisteria sinensis extract, 2-amino-3,5-dibrom-6-methylpyridine, pubchem7669
PubChem CID 522994
IUPAC Name 3,5-dibromo-6-methylpyridin-2-amine
SMILES CC1=NC(=C(C=C1Br)Br)N

Specifications

Melting Point 144°C to 145°C
Quantity 1 g
Beilstein 121839
Formula Weight 265.95
Percent Purity 97%
Chemical Name or Material 2-Amino-3,5-dibromo-6-methylpyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.