missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dibromobenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11361907
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-85-8
Molecular Formula C7H5Br2NO2
Molecular Weight (g/mol) 294.93
MDL Number MFCD00017087
InChI Key WNABMWFLKQEGCP-UHFFFAOYSA-N
Synonym 3,5-dibromoanthranilic acid, 2-amino-3,5-dibromobenzoicacid, 3,5-dibromo-2-aminobenzoic acid, buttpark 89\07-50, 2-amino-3,5-dibromo-benzoic acid, benzoic acid, 2-amino-3,5-dibromo, acmc-20ahts, pubchem3720, 2-carboxy-4,6-dibromoaniline, ghl.pd_mitscher_leg0.1128
PubChem CID 219491
IUPAC Name 2-amino-3,5-dibromobenzoic acid
SMILES NC1=C(Br)C=C(Br)C=C1C(O)=O

Specifications

Melting Point 230°C to 235°C
Quantity 5 g
Beilstein 779291
Formula Weight 294.93
Percent Purity 98%
Chemical Name or Material 2-Amino-3,5-dibromobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'