missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dimethylbenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11387086
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14438-32-5
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00017099
InChI Key GIMYRAQQQBFFFJ-UHFFFAOYSA-N
Synonym 3,5-dimethylanthranilic acid, 2-amino-3,5-dimethyl-benzoic acid, 2-amino-3,5-dimethyl benzoic acid, benzoic acid, 2-amino-3,5-dimethyl, pubchem4685, 3,5-dimethylanthranilic, acmc-1bqlm, intermediates-zcf02106, bidd:gt0362, 2-amino-3,5-dimethylbenzoicacid
PubChem CID 259834
IUPAC Name 2-amino-3,5-dimethylbenzoic acid
SMILES CC1=CC(=C(C(=C1)C(=O)O)N)C

Specifications

Melting Point 189°C to 194°C
Quantity 1 g
Beilstein 1101785
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material 2-Amino-3,5-dimethylbenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'