missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-chloropyridine, 95%, Thermo Scientific Chemicals

Product Code. 15453877
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39620-04-7
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD03541052
InChI Key RZJPBQGRCNJYBU-UHFFFAOYSA-N
Synonym 2-amino-3-chloropyridine, 3-chloro-pyridin-2-ylamine, 2-amino-3-chloro pyridine, 3-chloro-2-pyridinamine, 3-chloro-2-pyridinylamine, 2-pyridinamine, 3-chloro, 3-chloro-2-pyridylamine, 2-amino-3-chloro-pyridine, 3-chloranylpyridin-2-amine, o-aminochloropyridine
PubChem CID 693267
IUPAC Name 3-chloropyridin-2-amine
SMILES C1=CC(=C(N=C1)N)Cl

Specifications

Melting Point 59°C to 62°C
Quantity 250 mg
Formula Weight 128.56
Percent Purity 95%
Chemical Name or Material 2-Amino-3-chloropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'