missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(2-thienyl)benzoic acid, 96%, Thermo Scientific™

Product Code. 15456957
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 948006-04-0
Molecular Formula C11H9NO2S
Molecular Weight (g/mol) 219.258
MDL Number MFCD07357339
InChI Key RWXQPMJFVHLSST-UHFFFAOYSA-N
Synonym 2-amino-4-thiophen-2-yl benzoic acid, 2-amino-4-2-thienyl benzoic acid
PubChem CID 46741364
IUPAC Name 2-amino-4-thiophen-2-ylbenzoic acid
SMILES C1=CSC(=C1)C2=CC(=C(C=C2)C(=O)O)N

Specifications

Melting Point 229°C to 232°C
Quantity 250 mg
Formula Weight 219.26
Percent Purity 96%
Chemical Name or Material 2-Amino-4-(2-thienyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'