missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(2-thienyl)thiazole, 97%, Thermo Scientific™

Product Code. 15404408
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28989-50-6
Molecular Formula C7H6N2S2
Molecular Weight (g/mol) 182.259
MDL Number MFCD00457532
InChI Key XRNPDKQNHIKARG-UHFFFAOYSA-N
Synonym 4-thiophen-2-yl thiazol-2-amine, 4-thiophen-2-yl-thiazol-2-ylamine, 4-thiophen-2-yl-1,3-thiazol-2-amine, 4-2-thienyl-1,3-thiazol-2-amine, 2-amino-4-2-thienyl thiazole, 2-thiazolamine, 4-2-thienyl, 4-2-thienyl-1,3-thiazole-2-ylamine, ncimech_000023, ncistruc1_000175, ncistruc2_000001
PubChem CID 243973
IUPAC Name 4-thiophen-2-yl-1,3-thiazol-2-amine
SMILES C1=CSC(=C1)C2=CSC(=N2)N

Specifications

Color Brown
Quantity 2 g
Formula Weight 182.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-4-(2-thienyl)thiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'