missing translation for 'onlineSavingsMsg'
Learn More
2-Amino-4-(3-pyridyl)thiazole, 97%, Thermo Scientific™ Product Code.: 15494868

2-Amino-4-(3-pyridyl)thiazole, 97%, Thermo Scientific™

Product Code. 15494868
2 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 15494868

Brand: Thermo Scientific Alfa Aesar H58630.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30235-27-9
Molecular Formula C8H7N3S
Molecular Weight (g/mol) 177.225
MDL Number MFCD00218013
InChI Key XOHZQGAYUHOJPR-UHFFFAOYSA-N
Synonym 4-pyridin-3-yl thiazol-2-amine, 2-amino-4-3-pyridyl thiazole, 4-pyridin-3-yl-thiazol-2-ylamine, 4-pyridin-3-yl-1,3-thiazol-2-amine, 4-3-pyridinyl-2-thiazolamine, 2-thiazolamine, 4-3-pyridinyl, ccris 6699, 2-amino-4-pyridin-3-yl-1,3-thiazole, 4-pyridine-3-yl-thiazol-2-ylamine, 4-3-pyridinyl-1,3-thiazol-2-ylamineShow MoreShow Less
PubChem CID 154893
IUPAC Name 4-pyridin-3-yl-1,3-thiazol-2-amine
SMILES C1=CC(=CN=C1)C2=CSC(=N2)N

Specifications

Color Brown
Quantity 2 g
Formula Weight 177.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-4-(3-pyridyl)thiazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberH09Y029 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Amino-4-(3-pyridyl)thiazole, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.