missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(4-biphenylyl)thiazole, 97%, Thermo Scientific Chemicals

Product Code. 15440718
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2834-79-9
Molecular Formula C15H12N2S
Molecular Weight (g/mol) 252.335
MDL Number MFCD00047059
InChI Key HTAUVJPDFDVVHV-UHFFFAOYSA-N
Synonym 2-amino-4-4-biphenylyl thiazole, 4-biphenyl-4-yl-thiazol-2-ylamine, 4-4-phenylphenyl-1,3-thiazol-2-amine, usaf ek-4373, thiazole, 2-amino-4-4-biphenylyl, unii-39u8aa34ot, 4-1,1'-biphenyl-4-yl-1,3-thiazol-2-amine, 4-1,1'-biphenyl-4-yl thiazol-2-amine, 2-amino-4-4-biphenyl thiazole, 4-biphenyl-4-ylthiazol-2-ylamine
PubChem CID 76075
IUPAC Name 4-(4-phenylphenyl)-1,3-thiazol-2-amine
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CSC(=N3)N

Specifications

Quantity 5 g
Beilstein 187642
Formula Weight 252.33
Percent Purity 97%
Chemical Name or Material 2-Amino-4-(4-biphenylyl)thiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'