missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(5,6,7,8-tetrahydro-2-naphthyl)thiazole, 97%, Thermo Scientific™

Product Code. 15474136
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87999-04-0
Molecular Formula C13H14N2S
Molecular Weight (g/mol) 230.329
MDL Number MFCD01804671
InChI Key XCDNQJNJCJJQEC-UHFFFAOYSA-N
Synonym 4-5,6,7,8-tetrahydronaphthalen-2-yl-1,3-thiazol-2-amine, 2-amino-4-5,6,7,8-tetrahydro-2-naphthyl thiazole, 4-5,6,7,8-tetrahydro-naphthalen-2-yl-thiazol-2-ylamine, 4-5,6,7,8-tetrahydro-naphthalen-2-yl thiazol-2-ylamine, 4-1,2,3,4-tetrahydronaphthalen-6-yl thiazol-2-amine, 4-5,6,7,8-tetrahydronaphthalen-2-yl thiazol-2-ylamine, enamine_005054, 2-thiazolamine, 4-5,6,7,8-tetrahydro-2-naphthalenyl, 4-tetralin-6-yl thiazol-2-amine, 4-5,6,7,8-tetrahydronaphthalen-2-yl thiazol-2-amine
PubChem CID 735091
IUPAC Name 4-(5,6,7,8-tetrahydronaphthalen-2-yl)-1,3-thiazol-2-amine
SMILES C1CCC2=C(C1)C=CC(=C2)C3=CSC(=N3)N

Specifications

Melting Point 105°C to 110°C
Quantity 5 g
Sensitivity Light and moisture sensitive
Formula Weight 230.33
Percent Purity 97%
Chemical Name or Material 2-Amino-4-(5,6,7,8-tetrahydro-2-naphthyl)thiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.