missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4,6-dichloropyrimidine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Microwave-assisted synthesis of pyrazolo [3, 4-d] pyrimidines from 2-amino-4, 6-dichloropyrimidine-5-carbaldehyde is under solvent-free conditions. A series of 6-alkynyl-2,4-diaminopyrimidine derivatives bearing various substituents at alkynyl moiety was prepared by the Sonogashira cross-coupling reaction of 2,4-diamino-6-iodopyrimidine using Pd(PPh3)2Cl2 as catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Microwave-assisted synthesis of pyrazolo [3, 4-d] pyrimidines from 2-amino-4, 6-dichloropyrimidine-5-carbaldehyde is under solvent-free conditions. A series of 6-alkynyl-2,4-diaminopyrimidine derivatives bearing various substituents at alkynyl moiety was prepared by the Sonogashira cross-coupling reaction of 2,4-diamino-6-iodopyrimidine using Pd(PPh3)2Cl2 as catalyst.
Solubility
Insoluble in water, soluble in acetone, two monomer (VCM) and thermal toluene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Microwave-assisted synthesis of pyrazolo [3, 4-d] pyrimidines from 2-amino-4, 6-dichloropyrimidine-5-carbaldehyde is under solvent-free conditions. A series of 6-alkynyl-2,4-diaminopyrimidine derivatives bearing various substituents at alkynyl moiety was prepared by the Sonogashira cross-coupling reaction of 2,4-diamino-6-iodopyrimidine using Pd(PPh3)2Cl2 as catalyst.
Solubility
Insoluble in water, soluble in acetone, two monomer (VCM) and thermal toluene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
Melting Point | 218°C to 222°C |
Quantity | 5 g |
Beilstein | 118454 |
Solubility Information | Insoluble in water,soluble in acetone,two monomer (VCM) and thermal toluene. |
Formula Weight | 164 |
Percent Purity | 98% |
Chemical Name or Material | 2-Amino-4,6-dichloropyrimidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'