Learn More
2-Amino-4,6-dichloropyrimidine, 98%, Thermo Scientific Chemicals
£40.60 - £475.00
Chemical Identifiers
CAS | 56-05-3 |
---|---|
Molecular Formula | C4H3Cl2N3 |
Molecular Weight (g/mol) | 163.989 |
MDL Number | MFCD00006090 |
InChI Key | JPZOAVGMSDSWSW-UHFFFAOYSA-N |
Synonym | 2-amino-4,6-dichloropyrimidine, 4,6-dichloro-2-pyrimidinamine, 2-pyrimidinamine, 4,6-dichloro, 4,6-dichloropyrimidin-2-ylamine, ccris 7460, 4,6-dichloro-pyrimidin-2-ylamine, pyrimidine, 2-amino-4,6-dichloro, 2-amino-4,6-dichloro pyrimidine, 4,6-dichloropyrimidine-2-ylamine, zlchem 351 |
PubChem CID | 65522 |
IUPAC Name | 4,6-dichloropyrimidin-2-amine |
SMILES | C1=C(N=C(N=C1Cl)N)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11443734
|
Thermo Scientific Alfa Aesar
A11330.06 |
5 g |
£40.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11453734
|
Thermo Scientific Alfa Aesar
A11330.14 |
25 g |
£145.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399689
|
Thermo Scientific Alfa Aesar
A11330.22 |
100 g |
£475.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Microwave-assisted synthesis of pyrazolo [3, 4-d] pyrimidines from 2-amino-4, 6-dichloropyrimidine-5-carbaldehyde is under solvent-free conditions. A series of 6-alkynyl-2,4-diaminopyrimidine derivatives bearing various substituents at alkynyl moiety was prepared by the Sonogashira cross-coupling reaction of 2,4-diamino-6-iodopyrimidine using Pd(PPh3)2Cl2 as catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMicrowave-assisted synthesis of pyrazolo [3, 4-d] pyrimidines from 2-amino-4, 6-dichloropyrimidine-5-carbaldehyde is under solvent-free conditions. A series of 6-alkynyl-2,4-diaminopyrimidine derivatives bearing various substituents at alkynyl moiety was prepared by the Sonogashira cross-coupling reaction of 2,4-diamino-6-iodopyrimidine using Pd(PPh3)2Cl2 as catalyst.
Solubility
Insoluble in water, soluble in acetone, two monomer (VCM) and thermal toluene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
56-05-3 | |
163.989 | |
JPZOAVGMSDSWSW-UHFFFAOYSA-N | |
65522 | |
C1=C(N=C(N=C1Cl)N)Cl |
C4H3Cl2N3 | |
MFCD00006090 | |
2-amino-4,6-dichloropyrimidine, 4,6-dichloro-2-pyrimidinamine, 2-pyrimidinamine, 4,6-dichloro, 4,6-dichloropyrimidin-2-ylamine, ccris 7460, 4,6-dichloro-pyrimidin-2-ylamine, pyrimidine, 2-amino-4,6-dichloro, 2-amino-4,6-dichloro pyrimidine, 4,6-dichloropyrimidine-2-ylamine, zlchem 351 | |
4,6-dichloropyrimidin-2-amine |
Specifications
56-05-3 | |
C4H3Cl2N3 | |
118454 | |
Insoluble in water,soluble in acetone,two monomer (VCM) and thermal toluene. | |
C1=C(N=C(N=C1Cl)N)Cl | |
163.989 | |
164 | |
2-Amino-4,6-dichloropyrimidine |
218°C to 222°C | |
MFCD00006090 | |
2-amino-4,6-dichloropyrimidine, 4,6-dichloro-2-pyrimidinamine, 2-pyrimidinamine, 4,6-dichloro, 4,6-dichloropyrimidin-2-ylamine, ccris 7460, 4,6-dichloro-pyrimidin-2-ylamine, pyrimidine, 2-amino-4,6-dichloro, 2-amino-4,6-dichloro pyrimidine, 4,6-dichloropyrimidine-2-ylamine, zlchem 351 | |
JPZOAVGMSDSWSW-UHFFFAOYSA-N | |
4,6-dichloropyrimidin-2-amine | |
65522 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 200-253-9
missing translation for 'rtecsNumber' : UV6260485
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only