missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4,6-dihydroxypyrimidine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
2-Amino-4,6-dihydroxypyrimidine acts as an intermediate in the production of antimicrobial guanylsulfonamides. It also acts as an intermediate in pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-4,6-dihydroxypyrimidine acts as an intermediate in the production of antimicrobial guanylsulfonamides. It also acts as an intermediate in pharmaceuticals and agrochemicals.
Solubility
Soluble in Aqueous Alkali.
Notes
Store away from oxidizing agents. Store at -20°C. Store in a cool, dry, well-ventilated area.
2-Amino-4,6-dihydroxypyrimidine acts as an intermediate in the production of antimicrobial guanylsulfonamides. It also acts as an intermediate in pharmaceuticals and agrochemicals.
Solubility
Soluble in Aqueous Alkali.
Notes
Store away from oxidizing agents. Store at -20°C. Store in a cool, dry, well-ventilated area.
Specifications
Specifications
Melting Point | >300°C |
Quantity | 25 g |
Beilstein | 510297 |
Solubility Information | Soluble in Aqueous Alkali. |
Formula Weight | 127.1 |
Percent Purity | 98% |
Chemical Name or Material | 2-Amino-4,6-dihydroxypyrimidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'