missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-chloro-3-iodopyridine, 96%, Thermo Scientific™

£30.14 - £82.88

Chemical Identifiers

CAS 417721-69-8
Molecular Formula C5H4ClIN2
Molecular Weight (g/mol) 254.455
MDL Number MFCD08741521
InChI Key BWMULFKDCVPGNF-UHFFFAOYSA-N
Synonym 4-chloro-3-iodo-pyridin-2-ylamine, 2-amino-4-chloro-3-iodopyridine, 4-chloro-3-iodo-2-pyridinamine, pubchem14998, ksc585e3h, 4-chloro-3-iodo-2-pyridineamine, 4-chloro-3-iodopyridin-2-ylamine, 4-chloro-3-iodo-pyridin-2-yl amine
PubChem CID 22936596
IUPAC Name 4-chloro-3-iodopyridin-2-amine
SMILES C1=CN=C(C(=C1Cl)I)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15467037
View Documents
Thermo Scientific Alfa Aesar
H34118.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15457037
View Documents
Thermo Scientific Alfa Aesar
H34118.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

417721-69-8
254.455
BWMULFKDCVPGNF-UHFFFAOYSA-N
22936596
C1=CN=C(C(=C1Cl)I)N
C5H4ClIN2
MFCD08741521
4-chloro-3-iodo-pyridin-2-ylamine, 2-amino-4-chloro-3-iodopyridine, 4-chloro-3-iodo-2-pyridinamine, pubchem14998, ksc585e3h, 4-chloro-3-iodo-2-pyridineamine, 4-chloro-3-iodopyridin-2-ylamine, 4-chloro-3-iodo-pyridin-2-yl amine
4-chloro-3-iodopyridin-2-amine

Specifications

417721-69-8
C5H4ClIN2
Light sensitive
BWMULFKDCVPGNF-UHFFFAOYSA-N
4-chloro-3-iodopyridin-2-amine
22936596
97%
111°C to 113°C
MFCD08741521
4-chloro-3-iodo-pyridin-2-ylamine, 2-amino-4-chloro-3-iodopyridine, 4-chloro-3-iodo-2-pyridinamine, pubchem14998, ksc585e3h, 4-chloro-3-iodo-2-pyridineamine, 4-chloro-3-iodopyridin-2-ylamine, 4-chloro-3-iodo-pyridin-2-yl amine
C1=CN=C(C(=C1Cl)I)N
254.455
254.46
2-Amino-4-chloro-3-iodopyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.