missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-hydroxypyridine, 98%, Thermo Scientific Chemicals

Product Code. 15452219
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33631-05-9
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD04114161
InChI Key HQNIMNQVKVPZES-UHFFFAOYSA-N
Synonym 2-amino-4-hydroxypyridine, 2-aminopyridin-4-ol, 2-amino-4-pyridinol, 4-pyridinol, 2-amino, 2-amino-pyridin-4-ol, zlchem 31, pubchem6677, acmc-1acz5, 4-hydroxypyridine-2-amine, 2-aminopyridin-4 1h-one
PubChem CID 820936
IUPAC Name 2-amino-1H-pyridin-4-one
SMILES C1=CNC(=CC1=O)N

Specifications

Color Yellow
Quantity 1 g
Beilstein 385930
Solubility Information Slightly soluble in water.
Formula Weight 110.12
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Amino-4-hydroxypyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'