missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-methyl-5-nitropyridine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2-Amino-4-methyl-5-nitropyridine was used in the preparation of matrix mixture required to study new technical developments for the direct tissue analysis of peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-4-methyl-5-nitropyridine was used in the preparation of matrix mixture required to study new technical developments for the direct tissue analysis of peptides.
Solubility
Soluble in DMSO.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
2-Amino-4-methyl-5-nitropyridine was used in the preparation of matrix mixture required to study new technical developments for the direct tissue analysis of peptides.
Solubility
Soluble in DMSO.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications
Melting Point | 223°C to 225°C |
Quantity | 1 g |
Beilstein | 137695 |
Solubility Information | Soluble in DMSO. |
Formula Weight | 153.14 |
Percent Purity | 98% |
Chemical Name or Material | 2-Amino-4-methyl-5-nitropyridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'