missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester, 96%

Product Code. 15421116
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15421116

Brand: Thermo Scientific Alfa Aesar H54375.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 944401-55-2 | C11H18BN3O2 | 235.09 g/mol

2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 944401-55-2
Molecular Formula C11H18BN3O2
Molecular Weight (g/mol) 235.09
MDL Number MFCD18072554
InChI Key SYJMHOBGFXCKRG-UHFFFAOYSA-N
PubChem CID 52987906
IUPAC Name 4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
SMILES CC1=NC(N)=NC=C1B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 250 mg
Synonym 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 4-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-ylamine, 2-amino-4-methylpyrimidine-5-boronic acid pinacol ester, 4-methyl-2-aminopyrimidine-5-boronic acid pinacol ester, 2-amino-4-methylpyrimidin-5-yl boronic acid pinacol ester, 4-methyl-5-4, 4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl pyrimidine-2-ylamine, 4-methyl-5-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl pyrimidine-2-ylamine
Solubility Information Slightly soluble in water.
Formula Weight 235.09
Percent Purity 96%
Chemical Name or Material 2-Amino-4-methylpyrimidine-5-boronic acid pinacol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.