missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-methylthiazole, 98+%

Product Code. 11422627
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11422627

Brand: Thermo Scientific Alfa Aesar A14847.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1603-91-4 | C4H6N2S | 114.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1603-91-4
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.166
MDL Number MFCD00005329
InChI Key OUQMXTJYCAJLGO-UHFFFAOYSA-N
Synonym 2-amino-4-methylthiazole, 4-methylthiazol-2-amine, 2-thiazolamine, 4-methyl, 2-amino-4-methylthiazol, 4-methyl-2-thiazolamine, 4-methyl-2-thiazolylamine, 2-amino-4-methyl-1,3-thiazole, normotiroide, thiazole, 2-amino-4-methyl, 4-methyl-2-aminothiazole
PubChem CID 74143
ChEBI CHEBI:39753
IUPAC Name 4-methyl-1,3-thiazol-2-amine
SMILES CC1=CSC(=N1)N

Specifications

Melting Point 42°C to 46°C
Boiling Point 231°C to 233°C
Flash Point 107°C (224°F)
Odor Characteristic
Quantity 5 g
Beilstein 107890
Sensitivity Hygroscopic
Merck Index 14,450
Formula Weight 114.17
Percent Purity ≥98%
Chemical Name or Material 2-Amino-4-methylthiazole
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.