missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-nitrobenzoic acid, ≥97%, Thermo Scientific Chemicals

Product Code. 15482209
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-17-0
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.14
MDL Number MFCD00007262
InChI Key UEALKTCRMBVTFN-UHFFFAOYSA-N
Synonym 2-amino-4-nitrobenzoic acid, 4-nitroanthranilic acid, benzoic acid, 2-amino-4-nitro, 2-amino-4-nitrobenzoicacid, anthranilic acid, 4-nitro, ccris 441, 2-amino-4-nitro-benzoic acid, 4-nitro-2-aminobenzoic acid, dsstox_cid_963, dsstox_rid_75891
PubChem CID 12076
ChEBI CHEBI:75335
IUPAC Name 2-amino-4-nitrobenzoic acid
SMILES NC1=CC(=CC=C1C(O)=O)[N+]([O-])=O

Specifications

Melting Point 257°C (literature)
Quantity 500 g
Beilstein 785071
Formula Weight 182.13
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 2-Amino-4-nitrobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'