Learn More
2-Amino-5-bromo-4-(trifluoromethyl)pyridine, 95%, Thermo Scientific™
£43.00 - £149.00
Chemical Identifiers
CAS | 944401-56-3 |
---|---|
Molecular Formula | C6H4BrF3N2 |
Molecular Weight (g/mol) | 241.01 |
MDL Number | MFCD09864896 |
InChI Key | AEWYLVDLWQPJGW-UHFFFAOYSA-N |
Synonym | 5-bromo-4-trifluoromethyl pyridin-2-amine, 2-amino-5-bromo-4-trifluoromethyl pyridine, 5-bromo-4-trifluoromethyl-2-pyridylamine, 2-amino-5-bromo-4-trifluoromethylpyridine, acmc-209rru, 2-amino-4-trifluoromethyl-5-bromopyridine, 5-bromo-4-trifluoromethyl-2-pyridinamine, 5-bromo-4-trifluoromethyl-2-pyridinylamine, 5-bromanyl-4-trifluoromethyl pyridin-2-amine |
PubChem CID | 28875348 |
IUPAC Name | 5-bromo-4-(trifluoromethyl)pyridin-2-amine |
SMILES | NC1=NC=C(Br)C(=C1)C(F)(F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
944401-56-3 | |
241.01 | |
AEWYLVDLWQPJGW-UHFFFAOYSA-N | |
28875348 | |
NC1=NC=C(Br)C(=C1)C(F)(F)F |
C6H4BrF3N2 | |
MFCD09864896 | |
5-bromo-4-trifluoromethyl pyridin-2-amine, 2-amino-5-bromo-4-trifluoromethyl pyridine, 5-bromo-4-trifluoromethyl-2-pyridylamine, 2-amino-5-bromo-4-trifluoromethylpyridine, acmc-209rru, 2-amino-4-trifluoromethyl-5-bromopyridine, 5-bromo-4-trifluoromethyl-2-pyridinamine, 5-bromo-4-trifluoromethyl-2-pyridinylamine, 5-bromanyl-4-trifluoromethyl pyridin-2-amine | |
5-bromo-4-(trifluoromethyl)pyridin-2-amine |
Specifications
944401-56-3 | |
Orange to Yellow | |
MFCD09864896 | |
5-bromo-4-trifluoromethyl pyridin-2-amine, 2-amino-5-bromo-4-trifluoromethyl pyridine, 5-bromo-4-trifluoromethyl-2-pyridylamine, 2-amino-5-bromo-4-trifluoromethylpyridine, acmc-209rru, 2-amino-4-trifluoromethyl-5-bromopyridine, 5-bromo-4-trifluoromethyl-2-pyridinamine, 5-bromo-4-trifluoromethyl-2-pyridinylamine, 5-bromanyl-4-trifluoromethyl pyridin-2-amine | |
NC1=NC=C(Br)C(=C1)C(F)(F)F | |
241.01 | |
241.01 | |
Solid |
66°C to 75°C | |
C6H4BrF3N2 | |
15519034 | |
AEWYLVDLWQPJGW-UHFFFAOYSA-N | |
5-bromo-4-(trifluoromethyl)pyridin-2-amine | |
28875348 | |
95% | |
2-Amino-5-bromo-4-(trifluoromethyl)pyridine |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures