missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromobenzothiazole, 97%, Thermo Scientific Chemicals

Product Code. 15491468
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Amino-5-bromobenzothiazole is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-bromobenzothiazole is used as pharmaceutical intermediates

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20358-03-6
Molecular Formula C7H5BrN2S
Molecular Weight (g/mol) 229.095
MDL Number MFCD00577806
InChI Key ZPUJTWBWSOOMRP-UHFFFAOYSA-N
Synonym 2-amino-5-bromobenzothiazole, 5-bromobenzo d thiazol-2-amine, 5-bromo-benzothiazol-2-ylamine, 2-benzothiazolamine, 5-bromo, 5-bromo-2-benzothiazolamine, 2-amino-5-bromo-1,3-benzothiazole, pubchem21702, acmc-209f9a, 2-amino-5-bromo benzothiazole, 5-bromobenzothiazole-2-ylamine
PubChem CID 13775785
IUPAC Name 5-bromo-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1Br)N=C(S2)N

Specifications

Melting Point 194°C to 198°C
Color White to Brown
Quantity 25 g
Solubility Information Soluble in water.
Formula Weight 229.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-5-bromobenzothiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'