Learn More
2-Amino-5-bromobenzothiazole, 97%, Thermo Scientific Chemicals
£37.70 - £409.00
Chemical Identifiers
CAS | 20358-03-6 |
---|---|
Molecular Formula | C7H5BrN2S |
Molecular Weight (g/mol) | 229.095 |
MDL Number | MFCD00577806 |
InChI Key | ZPUJTWBWSOOMRP-UHFFFAOYSA-N |
Synonym | 2-amino-5-bromobenzothiazole, 5-bromobenzo d thiazol-2-amine, 5-bromo-benzothiazol-2-ylamine, 2-benzothiazolamine, 5-bromo, 5-bromo-2-benzothiazolamine, 2-amino-5-bromo-1,3-benzothiazole, pubchem21702, acmc-209f9a, 2-amino-5-bromo benzothiazole, 5-bromobenzothiazole-2-ylamine |
PubChem CID | 13775785 |
IUPAC Name | 5-bromo-1,3-benzothiazol-2-amine |
SMILES | C1=CC2=C(C=C1Br)N=C(S2)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15471468
|
Thermo Scientific Alfa Aesar
H55270.03 |
1 g |
£37.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15481468
|
Thermo Scientific Alfa Aesar
H55270.06 |
5 g | |||||||
15491468
|
Thermo Scientific Alfa Aesar
H55270.14 |
25 g | |||||||
Description
2-Amino-5-bromobenzothiazole is used as pharmaceutical intermediates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-5-bromobenzothiazole is used as pharmaceutical intermediates
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
20358-03-6 | |
229.095 | |
ZPUJTWBWSOOMRP-UHFFFAOYSA-N | |
13775785 | |
C1=CC2=C(C=C1Br)N=C(S2)N |
C7H5BrN2S | |
MFCD00577806 | |
2-amino-5-bromobenzothiazole, 5-bromobenzo d thiazol-2-amine, 5-bromo-benzothiazol-2-ylamine, 2-benzothiazolamine, 5-bromo, 5-bromo-2-benzothiazolamine, 2-amino-5-bromo-1,3-benzothiazole, pubchem21702, acmc-209f9a, 2-amino-5-bromo benzothiazole, 5-bromobenzothiazole-2-ylamine | |
5-bromo-1,3-benzothiazol-2-amine |
Specifications
20358-03-6 | |
White to Brown | |
MFCD00577806 | |
Soluble in water. | |
C1=CC2=C(C=C1Br)N=C(S2)N | |
229.095 | |
229.1 | |
Solid |
194°C to 198°C | |
C7H5BrN2S | |
2-amino-5-bromobenzothiazole, 5-bromobenzo d thiazol-2-amine, 5-bromo-benzothiazol-2-ylamine, 2-benzothiazolamine, 5-bromo, 5-bromo-2-benzothiazolamine, 2-amino-5-bromo-1,3-benzothiazole, pubchem21702, acmc-209f9a, 2-amino-5-bromo benzothiazole, 5-bromobenzothiazole-2-ylamine | |
ZPUJTWBWSOOMRP-UHFFFAOYSA-N | |
5-bromo-1,3-benzothiazol-2-amine | |
13775785 | |
97% | |
2-Amino-5-bromobenzothiazole |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only