missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromonicotinic acid, 95%, Thermo Scientific™

Catalog Number 10140372
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 52833-94-0
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C6H5BrN2O2
MDL Number MFCD01860227
Quantity 1 g
Synonym 2-amino-5-bromonicotinic acid, 2-amino-5-bromo-nicotinic acid, 3-pyridinecarboxylic acid, 2-amino-5-bromo, 2-amino-5-bromonicotonic acid, 2-amino-5-bromo-3-pyridinecarboxylic acid, 2-amino-5-bromo-pyridine-3-carboxylic acid, 2-amino-5-bromo nicotinic acid, zlchem 461, pubchem5708
InChI Key IEPDTLRHISNBLB-UHFFFAOYSA-N
SMILES C1=C(C(=NC=C1Br)N)C(=O)O
IUPAC Name 2-amino-5-bromopyridine-3-carboxylic acid
Molecular Weight (g/mol) 217
PubChem CID 737445
Formula Weight 217
Percent Purity 95%
Chemical Name or Material 2-Amino-5-bromonicotinic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-5-bromonicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'