missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-chlorobenzamide, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2-Amino-5-chlorobenzamide react with 2-Azido-benzoyl chloride to get 2-Azido-5'-chloro-2'-carboxamidobenzanilide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-5-chlorobenzamide react with 2-Azido-benzoyl chloride to get 2-Azido-5′-chloro-2′-carboxamidobenzanilide.
Solubility
Soluble in acetone.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-Amino-5-chlorobenzamide react with 2-Azido-benzoyl chloride to get 2-Azido-5′-chloro-2′-carboxamidobenzanilide.
Solubility
Soluble in acetone.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 169°C to 173°C |
Quantity | 1 g |
Beilstein | 743347 |
Solubility Information | Soluble in acetone. |
Formula Weight | 170.6 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Amino-5-chlorobenzamide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'