missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorothiazole hydrochloride, 97%

Product Code. 11397555
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11397555

Brand: Thermo Scientific Alfa Aesar L05707.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55506-37-1 | C3H4Cl2N2S | 171.04 g/mol

2-Amino-5-chlorothiazole hydrochloride is used as starting reagent in the synthesis of 2-chloro-6-methylimidazo[2,1-b]thiazole. It is used as reactant for preparation of biologically active thiazole derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-chlorothiazole hydrochloride is used as starting reagent in the synthesis of 2-chloro-6-methylimidazo[2,1-b]thiazole. It is used as reactant for preparation of biologically active thiazole derivatives.

Solubility
Soluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55506-37-1
Molecular Formula C3H4Cl2N2S
Molecular Weight (g/mol) 171.04
MDL Number MFCD00039006
InChI Key GTMGFQYVLSQTKP-UHFFFAOYSA-N
Synonym 2-amino-5-chlorothiazole hydrochloride, 5-chlorothiazol-2-amine hydrochloride, 5-chloro-1,3-thiazol-2-amine hydrochloride, 2-thiazolamine, 5-chloro-, monohydrochloride, 5-chloro-2-thiazolamine hydrochloride, 2-amino-5-chloro-1,3-thiazole hydrochloride, 5-chlorothiazolamine hydrochloride, thiazole, 2-amino-5-chloro-hydrochloride, pubchem23694, c3h3cln2s.hcl
PubChem CID 148743
IUPAC Name 5-chloro-1,3-thiazol-2-amine;hydrochloride
SMILES [H+].[Cl-].NC1=NC=C(Cl)S1

Specifications

Melting Point 168°C to 172°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 171.05
Percent Purity 97%
Chemical Name or Material 2-Amino-5-chlorothiazole hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.