missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
2-Amino-5-fluorobenzeneboronic acid pinacol ester is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-5-fluorobenzeneboronic acid pinacol ester is used as a pharmaceutical intermediate.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
2-Amino-5-fluorobenzeneboronic acid pinacol ester is used as a pharmaceutical intermediate.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 49°C to 50°C |
Quantity | 250 mg |
Formula Weight | 237.08 |
Percent Purity | 96% |
Chemical Name or Material | 2-Amino-5-fluorobenzeneboronic acid pinacol ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'