missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylthiazole, 98%, Thermo Scientific Chemicals

Product Code. 10530554
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7305-71-7
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.17
MDL Number MFCD00078317
InChI Key GUABFMPMKJGSBQ-UHFFFAOYSA-N
Synonym 2-amino-5-methylthiazole, 5-methylthiazol-2-amine, 2-thiazolamine, 5-methyl, 2-amino-5-methyl-thiazole, thiazole, 2-amino-5-methyl, 5-methyl-2-thiazolamine, 5-methyl-2-aminothiazole, unii-7rab78af2p, 2-amino-5-methyl thiazole, meloxicam related compound b
PubChem CID 351770
IUPAC Name 5-methyl-1,3-thiazol-2-amine
SMILES CC1=CN=C(S1)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 93.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 114.17
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-5-methylthiazole
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Amino-5-methylthiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.