missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-nitrobenzoic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
2-Amino-5-nitrobenzoic acid, is used as an intermediate for dyes and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-5-nitrobenzoic acid, is used as an intermediate for dyes and pharmaceuticals.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to strong oxidizing agents. Stable under ordinary conditions.
2-Amino-5-nitrobenzoic acid, is used as an intermediate for dyes and pharmaceuticals.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to strong oxidizing agents. Stable under ordinary conditions.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼270°C (decomposition) |
Quantity | 10 g |
Beilstein | 646219 |
Solubility Information | Insoluble in water. |
Formula Weight | 182.14 |
Percent Purity | 98% |
Chemical Name or Material | 2-Amino-5-nitrobenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'